accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COOOO

CAS RN 4196-89-8

IUPAC Name: 3-(benzoyloxy)-2,2-dimethylpropyl benzoate
Synonyms: 3-(benzoyloxy)-2,2-dimethylpropyl benzoate
Molecular Weight (g/mol): 312.37
Molecular Formula: C19H20O4
InChi Key: DYJIIMFHSZKBDY-UHFFFAOYSA-N
SMILES: CC(C)(COC(=O)C1=CC=CC=C1)COC(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results