Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neopentyl Glycol Dibenzoate 98.0+%, TCI America™
SDP

Catalog No. N057310G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N057310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N057310G Supplier TCI America Supplier No. N057310G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4196-89-8
Molecular Formula C19H20O4
Molecular Weight (g/mol) 312.365
MDL Number MFCD00020674
InChI Key DYJIIMFHSZKBDY-UHFFFAOYSA-N
Synonym neopentyl glycol dibenzoate, 2,2-dimethylpropane-1,3-diyl dibenzoate, unii-jbp9mr90cq, 2,2-dimethyl-1,3-propanediol dibenzoate, 1,3-propanediol, 2,2-dimethyl-, dibenzoate, jbp9mr90cq, 3-benzoyloxy-2,2-dimethylpropyl benzoate, 1,3-propanediol, 2,2-dimethyl-, 1,3-dibenzoate, neopentylglycoldibenzoate, 2,3-propanediol dibenzoate
PubChem CID 20169
IUPAC Name (3-benzoyloxy-2,2-dimethylpropyl) benzoate
SMILES CC(C)(COC(=O)C1=CC=CC=C1)COC(=O)C2=CC=CC=C2

Specifications

Melting Point 49°C
Color White
Boiling Point 271°C
Quantity 10 g
Formula Weight 312.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Neopentyl Glycol Dibenzoate
EINECSNumber (7)-0705
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.