Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neopentyl Glycol Dibenzoate, MP Biomedicals

Catalog No. ICN225336 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225336 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225336 Supplier MP Biomedicals, Inc Supplier No. 0522533683

Chemical Identifiers

CAS 4196-89-8
Molecular Formula C19H20O4
Molecular Weight (g/mol) 312.365
InChI Key DYJIIMFHSZKBDY-UHFFFAOYSA-N
Synonym neopentyl glycol dibenzoate, 2,2-dimethylpropane-1,3-diyl dibenzoate, unii-jbp9mr90cq, 2,2-dimethyl-1,3-propanediol dibenzoate, 1,3-propanediol, 2,2-dimethyl-, dibenzoate, jbp9mr90cq, 3-benzoyloxy-2,2-dimethylpropyl benzoate, 1,3-propanediol, 2,2-dimethyl-, 1,3-dibenzoate, neopentylglycoldibenzoate, 2,3-propanediol dibenzoate
PubChem CID 20169
IUPAC Name (3-benzoyloxy-2,2-dimethylpropyl) benzoate
SMILES CC(C)(COC(=O)C1=CC=CC=C1)COC(=O)C2=CC=CC=C2

Specifications

Quantity 100 g
Chemical Name or Material Neopentyl Glycol Dibenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.