accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4197-25-5

IUPAC Name: 2,2-dimethyl-6-{2-[4-(2-phenyldiazen-1-yl)naphthalen-1-yl]diazen-1-yl}-2,3-dihydro-1H-perimidine
Synonyms: sudan black B
Molecular Weight (g/mol): 456.55
Molecular Formula: C29H24N6
InChi Key: YCUVUDODLRLVIC-UHFFFAOYSA-N
SMILES: CC1(C)NC2=CC=C(N=NC3=C4C=CC=CC4=C(C=C3)N=NC3=CC=CC=C3)C3=CC=CC(N1)=C23
Molecular Weight (g/mol) 
  • (1)
  • (11)
Quantity 
  • (5)
  • (2)
  • (5)
Color 
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
Grade 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)
  • (1)
  • (11)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)