Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan Black B, MP Biomedicals

Catalog No. ICN206470 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN206470 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN206470 Supplier MP Biomedicals, Inc Supplier No. 0520647010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4197-25-5
Molecular Formula C29H24N6
Molecular Weight (g/mol) 456.553
InChI Key YCUVUDODLRLVIC-UHFFFAOYSA-N
PubChem CID 61336
IUPAC Name (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene
SMILES CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C

Specifications

Formula Weight 456.55
Quantity 10 g
Chemical Name or Material Sudan Black B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.