accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFF

CAS RN 421-53-4

IUPAC Name: 2,2,2-trifluoroacetaldehyde
Synonyms: trifluoroacetaldehyde
Molecular Weight (g/mol): 98.02
Molecular Formula: C2HF3O
InChi Key: JVTSHOJDBRTPHD-UHFFFAOYSA-N
SMILES: FC(F)(F)C=O
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)