Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoro-1,1-ethanediol (contains Total ca. 25% Water), TCI America™
SDP

Catalog No. T32795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T32795G Supplier TCI America Supplier No. T32795G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 421-53-4
Molecular Formula C2HF3O
Molecular Weight (g/mol) 98.02
MDL Number MFCD00042444
InChI Key JVTSHOJDBRTPHD-UHFFFAOYSA-N
Synonym tfaih, fluoral hydrate, 1,1-ethanediol, 2,2,2-trifluoro, fluoral, hydrate, 2,2,2-trifluoro-1,1-ethanediol, acetaldehyde, trifluoro-, hydrate, 2,2,2-trifluoroethanediol, fluoral hydrate;, snpljqhjptadtaeup@, acetaldehyde, hydrate
PubChem CID 67901
IUPAC Name 2,2,2-trifluoroacetaldehyde
SMILES FC(F)(F)C=O

Specifications

Color Colorless
Boiling Point 106°C
Quantity 5 g
UN Number 1989
Formula Weight 116.04
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoro-1,1-ethanediol (contains Total ca. 25% Water)
RTECSNumber KI2850000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.