Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetaldehyde hydrate, tech., ca 75% in water

Catalog No. AAA1323418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1323418 50 g
AAA1323409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1323418 Supplier Thermo Scientific Chemicals Supplier No. A1323418
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 421-53-4 | C2HF3O | 98.02 g/mol

One general application of trifluoroacetaldehyde is the preparation of trifluoroethylamino derivatives via reductive amination reaction. This synthesis includes the formation of the corresponding N,O-acetal intermediates followed by their reduction using NaBH4 or 2-picoline borane complex, affording the target trifluoroethylamino compounds in moderate to good yields. Another general application of trifluoroacetaldehyde is the synthesis of chiral α-substituted trifluoroethylamino compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
One general application of trifluoroacetaldehyde is the preparation of trifluoroethylamino derivatives via reductive amination reaction. This synthesis includes the formation of the corresponding N,O-acetal intermediates followed by their reduction using NaBH4 or 2-picoline borane complex, affording the target trifluoroethylamino compounds in moderate to good yields. Another general application of trifluoroacetaldehyde is the synthesis of chiral α-substituted trifluoroethylamino compounds.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Keep in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 421-53-4
Molecular Formula C2HF3O
Molecular Weight (g/mol) 98.02
MDL Number MFCD00042444
InChI Key JVTSHOJDBRTPHD-UHFFFAOYSA-N
Synonym tfaih, fluoral hydrate, 1,1-ethanediol, 2,2,2-trifluoro, fluoral, hydrate, 2,2,2-trifluoro-1,1-ethanediol, acetaldehyde, trifluoro-, hydrate, 2,2,2-trifluoroethanediol, fluoral hydrate;, snpljqhjptadtaeup@, acetaldehyde, hydrate
PubChem CID 67901
IUPAC Name 2,2,2-trifluoroethane-1,1-diol
SMILES FC(F)(F)C=O

Specifications

Density 1.44
Boiling Point 104°C to 106°C
Flash Point None
Linear Formula CF3CH(OH)2
Quantity 50 g
UN Number UN1989
Solubility Information Soluble in water.
Formula Weight 116.04 (Anhydrous)
Percent Purity 75%
Grade Technical
Concentration or Composition (by Analyte or Components) ≈75% in water
Chemical Name or Material Trifluoroacetaldehyde hydrate
Show More Show Less
Hazard Category H225-H302-H315-H317-H318-H335
Hazard Statement GHS H Statement
H225-H318-H302-H317-H335-H315
Highly flammable liquid and vapour.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 207-006-4
RTECSNumber KI2850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.