accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42149-74-6

IUPAC Name: 1-(2-chloroethoxy)propane
Synonyms: 1-(2-chloroethoxy)propane
Molecular Weight (g/mol): 122.59
Molecular Formula: C5H11ClO
InChi Key: BHDSGQOSIWVMJW-UHFFFAOYSA-N
SMILES: CCCOCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results