Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl Propyl Ether 97.0+%, TCI America™
SDP

Catalog No. C117425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C117425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C117425ML Supplier TCI America Supplier No. C117425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42149-74-6
Molecular Formula C5H11ClO
Molecular Weight (g/mol) 122.592
MDL Number MFCD00059052
InChI Key BHDSGQOSIWVMJW-UHFFFAOYSA-N
Synonym 2-Propoxyethyl Chloride
PubChem CID 123506
IUPAC Name 1-(2-chloroethoxy)propane
SMILES CCCOCCCl

Specifications

Color Yellow
Boiling Point 129°C
Quantity 25 mL
UN Number 3271
Formula Weight 122.59
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Propyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.