CAS RN 4221-03-8
IUPAC Name:
5-hydroxypentanal
Synonyms:
5-hydroxypentanal
Molecular Weight (g/mol):
102.13
Molecular Formula:
C5H10O2
InChi Key:
CNRGMQRNYAIBTN-UHFFFAOYSA-N
SMILES:
OCCCCC=O
1
–
1
of
1
results
1
R&D Systems™ (S)-(-)-Propranolol hydrochloride
CAS: 4221-03-8 Molecular Formula: C5H10O2