CAS RN 42252-34-6
CAS RN 42252-34-6
IUPAC Name:
N-ethyl-N-methylcarbamoyl chloride
Synonyms:
N-ethyl-N-methylcarbamoyl chloride
Molecular Weight (g/mol):
121.56
Molecular Formula:
C4H8ClNO
InChi Key:
XZVYDRLPXWFRIS-UHFFFAOYSA-N
SMILES:
CCN(C)C(Cl)=O
1
–
1
of
1
results
1
N-Ethyl-N-methylcarbamoyl Chloride 98.0+%, TCI America™
CAS: 42252-34-6 Molecular Formula: C4H8ClNO Molecular Weight (g/mol): 121.56 MDL Number: MFCD08061404 InChI Key: XZVYDRLPXWFRIS-UHFFFAOYSA-N PubChem CID: 11240471 IUPAC Name: N-ethyl-N-methylcarbamoyl chloride SMILES: CCN(C)C(Cl)=O