accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOOH(E/Z)

CAS RN 42438-89-1

IUPAC Name: 3-[2-(4-hydroxyphenyl)ethenyl]-5-methoxyphenol
Synonyms: pinostilbene
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: KUWZXOMQXYWKBS-UHFFFAOYSA-N
SMILES: COC1=CC(C=CC2=CC=C(O)C=C2)=CC(O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results