accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42461-84-7

IUPAC Name: (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol; 2-{[2-methyl-3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
Synonyms: flunixin; meglumine
Molecular Weight (g/mol): 491.46
Molecular Formula: C21H28F3N3O7
InChi Key: MGCCHNLNRBULBU-WZTVWXICSA-N
SMILES: CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.CC1=C(NC2=NC=CC=C2C(O)=O)C=CC=C1C(F)(F)F
Grade 
  • (1)
  • (6)
Percent Purity 
  • (6)
Physical Form 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (2)
  • (6)
  • (1)
  • (6)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)