Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flunixin Meglumine, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602756 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-756 10 kg Amber Glass Bottle
18-602-751 1 g Amber Glass Bottle
18-602-752 5 g Amber Glass Bottle
18-602-753 25 g Amber Glass Bottle
18-602-754 100 g Amber Glass Bottle
18-602-755 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-756 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F129310KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

F1293, 42461-84-7, C14H11F3N2O2·C7H17NO5

Spectrum™ Chemical Flunixin Meglumine, USP is a nonsteroidal, anti-inflammatory drug and antipyretic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 42461-84-7
Molecular Formula C21H28F3N3O7
Molecular Weight (g/mol) 491.46
InChI Key MGCCHNLNRBULBU-WZTVWXICSA-N
IUPAC Name (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol; 2-{[2-methyl-3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
SMILES CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.CC1=C(NC2=NC=CC=C2C(O)=O)C=CC=C1C(F)(F)F

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 0.5%
Melting Range 135°C to 140°C
Packaging Amber Glass Bottle
Quantity 10 kg
UN Number UN2811
Specific Rotation -9° to -12°
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Flunixin Meglumine, USP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.