accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFFFCl

CAS RN 425-87-6

IUPAC Name: 2-chloro-1,1,2-trifluoro-1-methoxyethane
Synonyms: 2-chloro-1,1,2-trifluoro-1-methoxyethane
Molecular Weight (g/mol): 148.51
Molecular Formula: C3H4ClF3O
InChi Key: KKXBMWAROXAWSZ-UHFFFAOYNA-N
SMILES: COC(F)(F)C(F)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results