Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,1,2-trifluoroethyl Methyl Ether 99.0+%, TCI America™
SDP

Catalog No. C08535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C08535G Supplier TCI America Supplier No. C08535G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 425-87-6
Molecular Formula C3H4ClF3O
Molecular Weight (g/mol) 148.509
MDL Number MFCD00039335
InChI Key KKXBMWAROXAWSZ-UHFFFAOYSA-N
PubChem CID 9877
IUPAC Name 2-chloro-1,1,2-trifluoro-1-methoxyethane
SMILES COC(C(F)Cl)(F)F

Specifications

Color Colorless
Boiling Point 70°C
Quantity 5 g
UN Number 1992
Formula Weight 148.51
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-1,1,2-trifluoroethyl Methyl Ether
RTECSNumber KN6825000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.