CAS RN 42523-29-5
CAS RN 42523-29-5
IUPAC Name:
2,7-dihydroxy-9H-fluoren-9-one
Synonyms:
2,7-dihydroxyfluoren-9-one
Molecular Weight (g/mol):
212.2
Molecular Formula:
C13H8O3
InChi Key:
CWHPQXRTQSNTRR-UHFFFAOYSA-N
SMILES:
OC1=CC2=C(C=C1)C1=C(C=C(O)C=C1)C2=O
1
–
1
of
1
results
1
2,7-Dihydroxy-9H-fluoren-9-one 98.0+%, TCI America™
CAS: 42523-29-5 Molecular Formula: C13H8O3 Molecular Weight (g/mol): 212.20 MDL Number: MFCD00037141 InChI Key: CWHPQXRTQSNTRR-UHFFFAOYSA-N PubChem CID: 4354181 IUPAC Name: 2,7-dihydroxy-9H-fluoren-9-one SMILES: OC1=CC2=C(C=C1)C1=C(C=C(O)C=C1)C2=O