accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 4265-27-4

IUPAC Name: 2-butyl-1-benzofuran
Synonyms: 2-butyl-1-benzofuran
Molecular Weight (g/mol): 174.24
Molecular Formula: C12H14O
InChi Key: OVJKFJDEVKABNF-UHFFFAOYSA-N
SMILES: CCCCC1=CC2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results