accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFNHONHNNHNNBrBrFFF

CAS RN 426834-69-7

IUPAC Name: 3-[3,5-bis(trifluoromethyl)phenyl]-1-[2,4-dibromo-6-(2H-1,2,3,4-tetrazol-5-yl)phenyl]urea
Synonyms: 3-[3,5-bis(trifluoromethyl)phenyl]-1-[2,4-dibromo-6-(2H-1,2,3,4-tetrazol-5-yl)phenyl]urea
Molecular Weight (g/mol): 574.08
Molecular Formula: C16H8Br2F6N6O
InChi Key: UZWJWROOLOOCPQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(NC(=O)NC2=C(Br)C=C(Br)C=C2C2=NNN=N2)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)