accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42779-10-2

H3CCH3SOH

CAS RN 42779-10-2

IUPAC Name: 2-[(2-methylpropyl)sulfanyl]ethan-1-ol
Synonyms: 2-[(2-methylpropyl)sulfanyl]ethanol
Molecular Weight (g/mol): 134.24
Molecular Formula: C6H14OS
InChi Key: PSCYOFJZGIAXOL-UHFFFAOYSA-N
SMILES: CC(C)CSCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results