accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNHOO

CAS RN 42856-71-3

IUPAC Name: 2-methylpyrrolidine-2-carboxylic acid
Synonyms: 2-methylpyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: LWHHAVWYGIBIEU-UHFFFAOYNA-N
SMILES: CC1(CCCN1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)