Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Methyl-L-proline 96.0+%, TCI America™
SDP

Catalog No. M20775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20775G 5 g
M20771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20775G Supplier TCI America Supplier No. M20775G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42856-71-3
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD01318647
InChI Key LWHHAVWYGIBIEU-LURJTMIESA-N
PubChem CID 6993664
IUPAC Name (2S)-2-methylpyrrolidine-2-carboxylic acid
SMILES CC1(CCCN1)C(=O)O

Specifications

Melting Point 310°C
Color White
Quantity 5 g
Formula Weight 129.16
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Methyl-L-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.