accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOONFFFSFFFOOOOOO(E/Z)

CAS RN 428816-48-2

IUPAC Name: [3,4,5-tris(benzoyloxy)-6-({[4-(trifluoromethyl)phenyl]imino}({[4-(trifluoromethyl)phenyl]methyl}sulfanyl)methoxy)oxan-2-yl]methyl benzoate
Synonyms: [3,4,5-tris(benzoyloxy)-6-({[4-(trifluoromethyl)phenyl]imino}({[4-(trifluoromethyl)phenyl]methyl}sulfanyl)methoxy)oxan-2-yl]methyl benzoate
Molecular Weight (g/mol): 957.89
Molecular Formula: C50H37F6NO10S
InChi Key: HTRJVXKRCWWVSR-UHFFFAOYNA-N
SMILES: FC(F)(F)C1=CC=C(CSC(OC2OC(COC(=O)C3=CC=CC=C3)C(OC(=O)C3=CC=CC=C3)C(OC(=O)C3=CC=CC=C3)C2OC(=O)C2=CC=CC=C2)=NC2=CC=C(C=C2)C(F)(F)F)C=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results