accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHOH

CAS RN 42890-76-6

IUPAC Name: butane-1,2,4-triol
Synonyms: 1,2,4-butanetriol
Molecular Weight (g/mol): 106.12
Molecular Formula: C4H10O3
InChi Key: ARXKVVRQIIOZGF-UHFFFAOYNA-N
SMILES: OCCC(O)CO
Concentration 
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results