Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1,2,4-Butanetriol, 95%, 99% e.e., Thermo Scientific™

Catalog No. AC339450010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC339450010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC339450010 Supplier Thermo Scientific Chemicals Supplier No. 339450010
Request Bulk or Custom Formats

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42890-76-6
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00063213
InChI Key ARXKVVRQIIOZGF-UHFFFAOYNA-N
Synonym s-1,2,4-butanetriol, s---1,2,4-butanetriol, s-butane-1,2,4-triol, 2s-butane-1,2,4-triol, s---1,2,4-trihydroxybutane, --1,2,4-butanetriol, 1,2,4-butanetriol, s, 1,2,4-butanetriol, 2s, --s-1,2,4-butanetriol, 2s-1,2,4-butanetriol
PubChem CID 640997
IUPAC Name butane-1,2,4-triol
SMILES OCCC(O)CO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Enantiomeric Excess 98.5% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 95%
Refractive Index 1.4740 to 1.4760 (20°C, 589nm)
Linear Formula HOCHCHCH(OH)CHOH
Quantity 1 g
Specific Rotation -27° to -33° (20°C, 589nm) (c=1, CH3OH)
Formula Weight 106.12
Percent Purity ≥94%
Physical Form Viscous Liquid
Chemical Name or Material (S)-1, 2, 4-Butanetriol, 95%, (99% e.e.)
Show More Show Less
Hazard Category MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.