accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4291-63-8

IUPAC Name: (2R,3S,5R)-5-(6-amino-2-chloro-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Synonyms: cladribine cladribina mylinax 2ClAdo leustatin cladribine product CdA chlorodeoxyadenosine CldAdo 2-chloro-deoxyadenosine
Molecular Weight (g/mol): 285.69
Molecular Formula: C10H12ClN5O3
InChi Key: PTOAARAWEBMLNO-KVQBGUIXSA-N
SMILES: NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC(Cl)=N1
Form 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results