Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cladribine 98.0+%, TCI America™
SDP

Catalog No. C249950MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C249950MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C249950MG Supplier TCI America Supplier No. C249950MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4291-63-8
Molecular Formula C10H12ClN5O3
Molecular Weight (g/mol) 285.688
MDL Number MFCD00153939
InChI Key PTOAARAWEBMLNO-KVQBGUIXSA-N
Synonym 2-Chloro-2′C-deoxyadenosine, 2-Chloro-9-(2-deoxy-beta-D-arabinofuranosyl)adenine, CdA
PubChem CID 20279
ChEBI CHEBI:567361
IUPAC Name (2R,3S,5R)-5-(6-amino-2-chloropurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
SMILES C1C(C(OC1N2C=NC3=C2N=C(N=C3N)Cl)CO)O

Specifications

Melting Point 220°C
Color White
Quantity 50 mg
Formula Weight 285.69
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cladribine
RTECSNumber AU7357560
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.