CAS RN 42923-77-3
IUPAC Name:
6-methoxy-1,2,3,4-tetrahydroisoquinoline
Synonyms:
6-methoxy-1,2,3,4-tetrahydroisoquinoline
Molecular Weight (g/mol):
163.22
Molecular Formula:
C10H13NO
InChi Key:
YYTAYINRPUJPNH-UHFFFAOYSA-N
SMILES:
COC1=CC=C2CNCCC2=C1