accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNH2N

CAS RN 43002-19-3

IUPAC Name: 2-(3-amino-1H-isoindol-1-ylidene)propanedinitrile
Synonyms: 2-(3-aminoisoindol-1-ylidene)propanedinitrile
Molecular Weight (g/mol): 194.2
Molecular Formula: C11H6N4
InChi Key: BDXWTIVCFJPDDL-UHFFFAOYSA-N
SMILES: NC1=NC(=C(C#N)C#N)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results