accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNHOH2NOHNOHOHO

CAS RN 4306-24-5

IUPAC Name: 2-[2-(2-aminoacetamido)-4-methylpentanamido]-3-(4-hydroxyphenyl)propanoic acid
Synonyms: 2-[2-(2-aminoacetamido)-4-methylpentanamido]-3-(4-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol): 351.4
Molecular Formula: C17H25N3O5
InChi Key: AFWYPMDMDYCKMD-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(=O)CN)C(=O)NC(CC1=CC=C(O)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results