accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3COH3COH3C

CAS RN 43083-12-1

IUPAC Name: 1,1,1-trimethoxybutane
Synonyms: 1,1,1-trimethoxybutane
Molecular Weight (g/mol): 148.2
Molecular Formula: C7H16O3
InChi Key: JAFMOTJMRSZOJE-UHFFFAOYSA-N
SMILES: CCCC(OC)(OC)OC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)