Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl Orthobutyrate 96.0+%, TCI America™
SDP

Catalog No. O01795ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01795ML 5 mL
O017925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01795ML Supplier TCI America Supplier No. O01795ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 43083-12-1
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.202
MDL Number MFCD00008480
InChI Key JAFMOTJMRSZOJE-UHFFFAOYSA-N
Synonym trimethyl orthobutyrate, trimethylorthobutyrate, butane, 1,1,1-trimethoxy, ortho-n-butyric acid trimethyl ester, orthobutyric acid trimethyl ester, orthobutyricacidtrimethylester, 1,1,1-trimethoxybutan, trimethy orthobutyrate, pubchem21070, acmc-1aqt6
PubChem CID 123512
IUPAC Name 1,1,1-trimethoxybutane
SMILES CCCC(OC)(OC)OC

Specifications

Color Yellow
Boiling Point 146°C
Quantity 5 mL
UN Number 3272
Formula Weight 148.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl Orthobutyrate
EINECSNumber (2)-3914
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.