accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHBrFFF

CAS RN 431-34-5

IUPAC Name: 3-bromo-1,1,1-trifluoropropan-2-ol
Synonyms: 3-bromo-1,1,1-trifluoropropan-2-ol
Molecular Weight (g/mol): 192.96
Molecular Formula: C3H4BrF3O
InChi Key: VBHIIZIQRDVGDH-UHFFFAOYNA-N
SMILES: OC(CBr)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)