accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 43146-97-0

IUPAC Name: 1-(undecyloxy)undecane
Synonyms: 1-(undecyloxy)undecane
Molecular Weight (g/mol): 326.61
Molecular Formula: C22H46O
InChi Key: TXYKVMGAIGVXFY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCOCCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results