Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecyl Ether, TCI America™
SDP

Catalog No. U005125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U005125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U005125G Supplier TCI America Supplier No. U005125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 43146-97-0
Molecular Formula C22H46O
Molecular Weight (g/mol) 326.609
MDL Number MFCD00059282
InChI Key TXYKVMGAIGVXFY-UHFFFAOYSA-N
Synonym Diundecyl Ether
PubChem CID 6451962
IUPAC Name 1-undecoxyundecane
SMILES CCCCCCCCCCCOCCCCCCCCCCC

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 326.61
Physical Form Crystals
Chemical Name or Material Undecyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.