accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3PH3CH3CH3CBr

CAS RN 4317-07-1

IUPAC Name: tetraethylphosphanium bromide
Synonyms: tetraethylphosphanium bromide
Molecular Weight (g/mol): 227.13
Molecular Formula: C8H20BrP
InChi Key: LIXPXSXEKKHIRR-UHFFFAOYSA-M
SMILES: [Br-].CC[P+](CC)(CC)CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results