accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 43229-80-7

IUPAC Name: (2E)-but-2-enedioic acid; bis(N-[2-hydroxy-5-(1-hydroxy-2-{[1-(4-methoxyphenyl)propan-2-yl]amino}ethyl)phenyl]formamide)
Synonyms: bis(formoterol); fumaric acid
Molecular Weight (g/mol): 804.89
Molecular Formula: C42H52N4O12
InChi Key: OBRNDARFFFHCGE-WXXKFALUNA-N
SMILES: OC(=O)\C=C\C(O)=O.COC1=CC=C(CC(C)NCC(O)C2=CC=C(O)C(NC=O)=C2)C=C1.COC1=CC=C(CC(C)NCC(O)C2=CC=C(O)C(NC=O)=C2)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results