accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOBr

CAS RN 435283-95-7

IUPAC Name: 4-[2-(3-bromophenoxy)ethyl]morpholine
Synonyms: 4-[2-(3-bromophenoxy)ethyl]morpholine
Molecular Weight (g/mol): 286.17
Molecular Formula: C12H16BrNO2
InChi Key: YZKFMBAOEVINLP-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(OCCN2CCOCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results