accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4359-46-0

IUPAC Name: 2-ethyl-4-methyl-1,3-dioxolane
Synonyms: 2-ethyl-4-methyl-1,3-dioxolane
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: CSZCLQLJVFLXLI-UHFFFAOYNA-N
SMILES: CCC1OCC(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)