Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-4-methyl-1,3-dioxolane solution in acetonitrile (1000mg/L), Thermo Scientific™

Catalog No. AAH31933AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31933AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH31933AE Supplier Thermo Scientific Chemicals Supplier No. H31933AE

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4359-46-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD16622267
InChI Key CSZCLQLJVFLXLI-UHFFFAOYSA-N
Synonym 1,3-dioxolane, 2-ethyl-4-methyl, 1,3-dioxolane,2-ethyl-4-methyl, cis, 1,2-propanediol acetal, 1,3-dioxolane, 2-ethyl-4-methyl-, cis, cis-2-thyl-4-methyl-1,3-dioxolane, 2-ethyl-4-methyl-1,3-dioxolane, cis + trans, solution in acetonitrile 1000mg/l
PubChem CID 92210
IUPAC Name 2-ethyl-4-methyl-1,3-dioxolane
SMILES CCC1OCC(O1)C

Specifications

Quantity 100 mL
UN Number UN1648
Concentration or Composition (by Analyte or Components) 1000mg/L
Chemical Name or Material 2-Ethyl-4-methyl-1,3-dioxolane solution in acetonitrile
Hazard Category H225-H302+H312+H332-H319
Hazard Statement GHS H Statement
H225-H312-H332-H319
Highly flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONITRILE
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.