accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNN

CAS RN 4392-85-2

IUPAC Name: [2,2'-bipyridine]-6-carbonitrile
Synonyms: [2,2'-bipyridine]-6-carbonitrile
Molecular Weight (g/mol): 181.2
Molecular Formula: C11H7N3
InChi Key: QLZHLNAALBJVOE-UHFFFAOYSA-N
SMILES: N#CC1=NC(=CC=C1)C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results