accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4393-16-2

IUPAC Name: (4-methanesulfonylphenyl)methanaminium
Synonyms: (4-methanesulfonylphenyl)methanaminium
Molecular Weight (g/mol): 186.25
Molecular Formula: C8H12NO2S
InChi Key: VMNXLLDFGVEBLE-UHFFFAOYSA-O
SMILES: CS(=O)(=O)C1=CC=C(C[NH3+])C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results