Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylsulphonylbenzylamine hydrochloride, 95%

Catalog No. AC424950025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424950025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC424950025 Supplier Thermo Scientific Chemicals Supplier No. 424950025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4393-16-2 | C8H11NO2S | 185.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4393-16-2
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.241
MDL Number MFCD00216491
InChI Key VMNXLLDFGVEBLE-UHFFFAOYSA-N
Synonym 4-methylsulfonyl benzylamine, 4-methanesulfonylphenyl methanamine, 4-methylsulfonyl phenyl methanamine, 4-methylsulfonylbenzylamine, benzenemethanamine, 4-methylsulfonyl, unii-61o466wzw2, 4-methanesulfonyl benzylamine, 4-methylsulfonyl benzenemethanamine, 4-methanesulfonylphenyl methylamine, 4-methanesulfonyl-benzylamine
PubChem CID 57421
IUPAC Name (4-methylsulfonylphenyl)methanamine
SMILES CS(=O)(=O)C1=CC=C(C=C1)CN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 269°C to 277°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula H2NH2CC6H4SO2CH3·HCl
Quantity 2.5 g
Formula Weight 221.71
Percent Purity ≥94% (Acid)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Methylsulphonylbenzylamine hydrochloride, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.