accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNHNONN

CAS RN 440100-64-1

IUPAC Name: 1-(2-{[4-methyl-1-(pyrimidin-2-yl)piperidin-4-yl]amino}acetyl)pyrrolidine-2-carbonitrile
Synonyms: 1-(2-{[4-methyl-1-(pyrimidin-2-yl)piperidin-4-yl]amino}acetyl)pyrrolidine-2-carbonitrile
Molecular Weight (g/mol): 328.42
Molecular Formula: C17H24N6O
InChi Key: JERPHRNGJBSFAP-UHFFFAOYNA-N
SMILES: CC1(CCN(CC1)C1=NC=CC=N1)NCC(=O)N1CCCC1C#N
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results