Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K 579, Tocris Bioscience™

Catalog No. 279050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-905-0 50 mg
27-901-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-905-0 Supplier R&D Systems Supplier No. 2790/50
This item is not returnable. View return policy

Dipeptidyl peptidase IV (DPP-IV) inhibitor

Dipeptidyl peptidase IV inhibitor. Inhibits rat, canine, human and monkey enzymes with IC50 values of 3, 5, 8, and 8 nM respectively. Long-acting hypoglycemic agent; attenuates glucose excursion following glucose loading in Zucker fatty rats.

Chemical Identifiers

CAS 440100-64-1
Molecular Formula C17H24N6O
Molecular Weight (g/mol) 328.42
InChI Key JERPHRNGJBSFAP-AWEZNQCLSA-N
Synonym unii-74p4vv90ru, dipeptidylpeptidase iv inhibitor iv, k579, 2s-1-4-methyl-1-2-pyrimidinyl-4-piperidinyl amino acetyl-2-pyrrolidinecarbonitrile, 2-pyrrolidinecarbonitrile, 1-4-methyl-1-2-pyrimidinyl-4-piperidinyl amino acetyl, 2-pyrrolidinecarbonitrile, 1-2-4-methyl-1-2-pyrimidinyl-4-piperidinyl amino acetyl-, 2s, s-1-4-methyl-1-pyrimidin-2-yl piperidin-4-yl glycyl pyrrolidine-2-carbonitrile
PubChem CID 9818824
IUPAC Name (2S)-1-[2-[(4-methyl-1-pyrimidin-2-ylpiperidin-4-yl)amino]acetyl]pyrrolidine-2-carbonitrile
SMILES CC1(CCN(CC1)C2=NC=CC=N2)NCC(=O)N3CCCC3C#N

Specifications

Quantity 50 mg
Formula Weight 328.41
Percent Purity >99%
Chemical Name or Material K 579
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.