accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CN

CAS RN 4411-80-7

IUPAC Name: 6,6'-dimethyl-2,2'-bipyridine
Synonyms: 6,6'-dimethyl-2,2'-bipyridine
Molecular Weight (g/mol): 184.24
Molecular Formula: C12H12N2
InChi Key: OHJPGUSXUGHOGE-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=N1)C1=CC=CC(C)=N1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)