accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4442-41-5

IUPAC Name: (phenoxymethoxy)benzene
Synonyms: (phenoxymethoxy)benzene
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: VTXLTXPNXYLCQD-UHFFFAOYSA-N
SMILES: C(OC1=CC=CC=C1)OC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results