Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenoxymethane 98.0+%, TCI America™
SDP

Catalog No. D23525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23525G 5 g
D235225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23525G Supplier TCI America Supplier No. D23525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4442-41-5
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00191642
InChI Key VTXLTXPNXYLCQD-UHFFFAOYSA-N
Synonym Formaldehyde Diphenyl Acetal, Methylene Glycol Diphenyl Ether
PubChem CID 78183
IUPAC Name phenoxymethoxybenzene
SMILES C1=CC=C(C=C1)OCOC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 200.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diphenoxymethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.