accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4443-48-5

IUPAC Name: 2-bromo-1,7,7-trimethylbicyclo[2.2.1]heptane
Synonyms: 2-bromo-1,7,7-trimethylbicyclo[2.2.1]heptane
Molecular Weight (g/mol): 217.15
Molecular Formula: C10H17Br
InChi Key: OTOQMOVZIUGCQE-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C(Br)C2
Boiling Point 
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)